5-Hydroxy-1-tetralone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one

A132877-1G A132877-250MG A132877-100G A132877-25G A132877-10G A132877-5G
76646-980EA 23 USD
76646-980 76646-982 77615-064 77615-066 77747-947 77747-948
5-Hydroxy-1-tetralone 98%
5-Hydroxy-1-tetralone

Caution: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00001693
CAS Number: 28315-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR