Aftin-4 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N6-Methyl-(R)-roscovitine, (R)-2-((6-(Benzyl(methyl)amino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol
A107870-10MG
A107870-1MG
A107870-25MG
A107870-5MG
76646-804EA
101
USD
76646-804
76646-806
76646-808
76646-810
Aftin-4 97%
Aftin-4
Caution: for Research Use Only
Formula:
C₂₀H₂₈N₆O |
MDL Number:
MFCD30187584 CAS Number: 866893-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...