Aftin-4 97%

Supplier: Ambeed
Warning

Synonyms: N6-Methyl-(R)-roscovitine, (R)-2-((6-(Benzyl(methyl)amino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol

A107870-10MG A107870-1MG A107870-25MG A107870-5MG
76646-804EA 101 USD
76646-804 76646-806 76646-808 76646-810
Aftin-4 97%
Aftin-4

Caution: for Research Use Only

Formula: C₂₀H₂₈N₆O
MDL Number: MFCD30187584
CAS Number: 866893-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR