Oligo Pro II System
Supplier: AGILENT TECHNOLOGIES, INC
M5340AA
76645-398EA 204161.4 USD
76645-398
Oligo Pro II System
Electrophoresis Systems Capillary Electrophoresis Systems
Oligonucleotide purity analysis using parallel capillary electrophoresis with UV detection.
  • Fast separations - complete 12, 24, or 96 samples in about 1 hour
  • Improve product quality - monitor synthesis performance to demonstrate purity of ssDNA or ssRNA
  • Wide sizing range - achieve single base resolution through 60-mers
  • Throughput for all labs - variable throughput with the option of 12, 24, or 96 capillary arrays
  • Direct detection of oligos - no dye labeling needed, uses UV detection
  • Simplified report generation - advanced peak integration software and data reporting capabilities
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
The Agilent Oligo Pro II system delivers accurate purity assessments of ssDNA and ssRNA oligonucleotides without the use of dyes. Ensure optimum downstream performance of oligonucleotides up to 60-mer in length with n-1 resolution.
With 12-, 24-, and 96-capillary arrays available along with three 96-well plate sample drawers, the Oligo Pro II system quickly adjusts to meet changing throughput demands. Intuitive and automated operation simplify the measurement of oligonucleotide purity and help you focus on results.
Caution: For research use only. Not for use in diagnostic procedures.