Fragment Analyzer System, 5300
Supplier: AGILENT TECHNOLOGIES, INC
M5311AA
76645-282EA 94346.56 USD
76645-282
Fragment Analyzer System, 5300
Electrophoresis Systems Capillary Electrophoresis Systems
Parallel capillary electrophoresis system used for nucleic acid QC in medium to high-throughput labs.
  • Separate 48 or 96 samples in parallel with a single simple dilution step and easy instrument set up
  • Reduce sample analysis time by using batch processes features post electrophoresis to automatically generate results reports
  • Load and separate up to 288 samples for unattended and automated analysis of large numbers of samples
  • Easily adapts to changes in your workflow with flexible option like maximizing your runs or speed or resolution of individual samples
More Product Information