SCIEX 7600 QTOF with Zeno Trap and EAD System
Supplier: SCIEX
5080337
76644-998EA 954635.35 USD
76644-998
SCIEX 7600 QTOF with Zeno Trap and EAD System
Spectrophotometers and Photometers Mass Spectrometers
This high-resolution accurate mass system combines Zeno trap pulsing with EAD fragmentation (electron activated dissociation) to unlock sensitivity gains that reveal new, rare, or even previously undetected information. Detect up to 20× more ions in every experiment and access a spectrum of tunable fragmentation techniques to uncover new perspectives.
  • Characterize large molecules including post-translational modifications
  • Elucidate positional isomers on small molecules and lipids
  • Identify and quantify proteins and peptides at unparalleled speed
  • Gain significant sensitivity and discover less abundant ions
  • EAD cell-tunable EAD between 0 to 25 eV
  • Source: Turbo V ion source housing with Twinspray analytical probe for ESI or APCI; OptiFlow ion source housing with Optiflow micro and nano probes
A high-resolution mass spectrometry solution that combines powerful MS/MS sensitivity, fragmentation technology and a step-change in data independent acquisition.