SCIEX X500B QTOF System
Supplier: SCIEX
5049811
76644-984EA 498807.86 USD
76644-984
SCIEX X500B QTOF System
Spectrophotometers and Photometers Mass Spectrometers
Engineered for standard biotherapeutic characterization analyzes, the X500B QTOF system is a true benchtop instrument, with streamlined processing to get to a consistent, accurate view of your biologic. It is optimized for intact mass analysis, subunit analysis as well as high-quality peptide mapping workflows.
  • Novel N-optic TOF design gives optimal mass accuracy and resolution, even for large biotherapeutics such as mAbs and antibody drug conjugates
  • Renowned turbo V ion source provides efficient ion production while limiting contamination for maximized robustness and uptime
  • Purpose built for biologics characterization and engineered with ease-of-use and simplicity in mind to accelerate and streamline biotherapeutic analysis
  • Perform intact mass, subunit analysis, and peptide mapping characterization on this single, true benchtop system
  • Source: Turbo V ion source housing with Twinspray analytical probe for ESI or APCI
And with tools to enable a complete Multiple Attribute Methodology (MAM) workflow, to allow you to define, track and quantify product quality attributes as well as specified contaminants, or find new unspecified impurities in a single software solution.