SCIEX X500R QTOF System
Supplier: SCIEX
5045633
76644-982EA 436492.71 USD
76644-982
SCIEX X500R QTOF System
Spectrophotometers and Photometers Mass Spectrometers
Engineered and versatile to handle the demands of any high throughput laboratory. Incorporating the robustness of turbo V ionization source along with MRMHR, it quantifies food and environmental testing samples to regulatory compliance requirements. For complete unknown screening, SWATH® acquisition detects any trace level compounds and delivers vital MS/MS data.
  • Renowned SWATH Data Independent Acquisition (DIA) enables comprehensive detection, so you won’t miss any critical MS/MS information for your application needs
  • SCIEX spectral libraries offer more compounds for more complete screening studies
  • Gain efficiency, see improved usability and achieve greater integrity with easy to learn and use SCIEX OS software
  • Source: TurboV ion source housing with Twinspray analytical probe for ESI or APCI
With simple workflows, robust hardware, sensitivity, and precision, the X500R QTOF is the first high-resolution LC-MS/MS system designed for both characterization and quantitation.