Tissue Cassette Storage Cabinet for Histology, Four E's USA
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
TCS100101
76644-974EA
206.46
USD
76644-974
Tissue Cassette Storage Cabinet for Histology, Four E's USA
Furniture
Storage Cabinets
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...