Advanced Slide Warmer
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
SDR100101
76644-948EA 2307.45 USD
76644-948
Advanced Slide Warmer
Slide Warmers/Dryers
Advanced slide dryer with angled slide placement area and touch screen.
  • Accurate temperature control
  • Angled slide placement area for ease of use
  • Intuitive touch screen control
  • Quiet, fan less design
  • High capacity slide drying or warming
Advanced slide dryer has a timer and auto on/off feature.
Heat up time: 20 min
Auto on/off: Yes
Delivery information: Typically in stock, ready to ship.
Packaging: Shipped in sturdy cardboard box with foam packaging.