Locking Incubator/Contrast Media Warmer
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
CMW100101
76644-934EA 1223.03 USD
76644-934
Locking Incubator/Contrast Media Warmer
Incubators Compact Benchtop Incubators
18 Liter locking incubator with digital lock, digital temperature control and timer.
  • Accurate temperature control with easy to use digital control panel
  • Durable cold rolled epoxy painted steel construction
  • Insulated glass viewing window for improved visibility
  • Built-in digital timer for managing timed processes samples
  • Digital lock for securing samples without the need for a key
Large handle and viewing window. Includes two shelves, stainless steel interior.
Heater type: Long life nickel alloy
Temperature control method: PID digital controller
Door lock: Digital keypad
Temperature control range: RT+5 to 65 °C
Delivery information: Typically in stock, ready to ship.
Packaging: Shipped in sturdy cardboard box with foam packaging.