Clinical Centrifuge, Four E's USA
Supplier: FOUR E#S INDUSTRIAL GROUP INC.
CFG300100
76644-932EA 1156.47 USD
76644-932
Clinical Centrifuge, Four E's USA
Centrifuges Benchtop Centrifuges
Four E's is a leading manufacturer of small centrifuges for clinical use.
  • Brushless DC motor for quiet operation
  • Easy to use digital control interface
  • Built-in safety features
  • Accommodates most laboratory centrifuge tubes
  • Control increment: 100 rpm
Four E's manufacturers a variety of centrifuges for clinical and research applications. The clinical centrifuge is the right size for small and medium size clinical laboratories that use small to medium size microcentrifuge tubes. The small footprint and the easy to use interface makes it perfect for dedicated testing use. The ultra-quiet brushless DC motor will keep this unit running for years to come.
Certifications: UL/CSA Power Supply, CE, RoHS.