VWR® Plus Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-730 76644-734 76644-738
76644-730EA 7237.88 USD
76644-730 76644-734 76644-738
VWR® Plus Manual Defrost Freezers with Inner Doors
Freezers Laboratory / General Purpose Freezers
The inner doors on these Plus Series freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-730, 76644-734, and 76644-738
Ordering information: All units come with a two-year parts and labor warranty and a five-year compressor parts warranty.