Transport Swabs without Tube
Supplier: Hardy Diagnostics
4175BX
76642-516PK 245.84 USD
76642-516
Transport Swabs without Tube
Swabs Transport Swabs
A sterile, single dacron swab on a straight aluminum wire individually wrapped in a peel pouch without media.
  • Aluminum wire with dacron tip
  • Individually wrapped
  • Sterile
  • No media
For ear, nose, throat, eye, and urethral use.