Cole-Parmer® DC-400 Automatic Desiccator Cabinet with Gas Purge Port, Antylia Scientific
Supplier: Antyila Scientific
78902-86
76640-526EA 2380.43 USD
76640-526
Cole-Parmer® DC-400 Automatic Desiccator Cabinet with Gas Purge Port, Antylia Scientific
Desiccators Desiccator Cabinets
Gas purge port for applications requiring a dry inert gas.
  • Nitrogen flowmeter allows humidity control from 3 to 20% RH
  • Built-in digital hygrometer allows easy monitoring of RH
  • View samples through acrylic viewing panels
Keep the moisture level very low using a desiccator with a gas purge port. Gas purge port allows for the flow of dry inert gas to further reduce the humidity in the cabinet—ideal for moisture-proof and anti-oxidation storage applications.
The electronic dehumidification system includes a permanent desiccant and a hygrometer to keep the relative humidity level at a constant level. The desiccant automatically recycles by discharging absorbed moisture through an external shutter. Once it is recycled, desiccant is ready to absorb moisture again.
Aluminum frame and stainless steel shelf offer excellent durability. Stainless steel shelf has holes to ensure uniform humidity throughout chamber. A lock and key allow you to securely lock the cabinet to prevent unauthorized access.
Delivery information: Supplied with one stainless steel shelf and a key.