Cole-Parmer® Essentials Disposable Forceps, Antylia Scientific
Supplier: Antyila Scientific
06443-20
76640-518PK 129.38 USD
76640-518
Cole-Parmer® Essentials Disposable Forceps, Antylia Scientific
Forceps
Get a more precise grip.
  • Corrugated handles provide secure grip
  • Reduces cross contamination
  • Disposable
  • Individually wrapped
Reduce cross contamination with these disposable forceps. Corrugated handles provide you with a secure grip. Made from polystyrene.
Sterile forceps are individually wrapped with easy to open peel-packs. Serrated blunt tips are more for general use.