AimStrip® Fentanyl Test Strips, Germaine Laboratories
Supplier: Germaine Laboratories
43345
76640-386PK 33.14 USD
76640-386
AimStrip® Fentanyl Test Strips, Germaine Laboratories
Drug Tests
AimStrip® Fentanyl Test Strips are used for the detection of Fentanyl in liquids and powders.
  • 25 Strips/Kit
  • Sensitivity 10 ng/mL
  • Results in 5 minutes
Packaging: Made in USA.
Caution: For Forensic Use Only.