Corning® Laboratory Bottles, Low Actinic, Ace Glass
Supplier: Ace Glass
4049-04 4049-06 4049-08 4049-10 4049-12 4049-18 4049-21 4049-25 4049-34
76529-606CS 176.57 USD
76529-606 76529-608 76529-610 76529-612 76529-614 76529-616 76529-618 76529-620 76529-622 76638-904 76638-906 76638-898 76638-902 76638-910 76638-896 76638-900 76638-908 76638-894
Corning® Laboratory Bottles, Low Actinic, Ace Glass
Bottles Glass Bottles
Media storage bottles with low actinic stain fired into the exterior surface.
  • Heavy-duty bottle of low actinic glass
  • Excellent for storing/transporting light senstive materials
  • Autoclavable
Protect materials sensitive to light in the 200 to 500 nanometer range. Glass bead indicates full capacity line. Permanent white enamel graduations and marking spots included.
Delivery information: Includes a linerless, one-piece, autoclavable (120 °C), orange polypropylene, plug-seal cap with drip-free pouring rings.