Pierce Plate, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0398
76638-832EA 558.69 USD
76638-832
Pierce Plate, Azenta Life Sciences
Microplates Deep Well Microplates
A metal block with 96 pins, suitable for piercing every well of a heat or adhesive sealed 96 well plate.
  • Machine-engineered metal block with 96 pins aligned central to each well of a 96 well plate
  • Pierce every well of a heat or adhesivesealed 96 well PCR or microplate (pierceable seals only)
  • Enables instant access to samples with a single or multichannel pipette or automated system
Clean between uses to avoid contamination by using most cleaning agents that are suitable for use on aluminium e.g. RNase removal solutions, bleach or UV treatment.