White Marker Pen
Supplier: AZENTA US, INC
4TI-0394
76638-826EA 19.8 USD
76638-826
White Marker Pen
Pens Lab Markers
This white marker pen is ideal for labeling Optically Clear Tissue Culture Plates, FrameStar® plates, 8 Well PCR Tube Strips with PC Frame, and other colored microplates or PCR plates.
  • White ink ensures clear labeling details
  • Ideal for labeling colored microplates or PCR plates
  • Ink is resistant to aqueous or organic solvents
The resulting waterproof ink cannot be removed using aqueous or organic solvents.
Packaging: One pen per case.