Choice® SatPax 500 Pre-wetted 60% Cellulose/40% Polyester Nonwoven Cleanroom Wipers, Berkshire
Supplier: Berkshire
SPXCHN50018
76638-266CS 257.25 USD
76638-266
Choice® SatPax 500 Pre-wetted 60% Cellulose/40% Polyester Nonwoven Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Choice® SatPax 500 wipes are composed of a hydroentangled nonwoven blend of 60% cellulose and 40% polyester. This pre-wetted format provides a cost-effective and easy-to-use solution versus traditional bulk handling of solvents.
  • Excellent combination of synthetic polyester strength and cleanliness, with the absorbent characteristics of cellulose
  • Smooth and durable with good wet strength
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Reduces VOC emissions
Choice® value tiered category of wipers offers all the performance consistency of higher priced wipers. Choice® 500 wipes are composed of a hydroentangled nonwoven blend of 60% cellulose and 40% polyester, with three saturation levels of isopropyl alcohol and DI water. This pre-wetted format provides a cost-effective and easy-to-use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with prepping dry wipers.
Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100,000. EU Grade B/C/D.