Storage Microplates, 96 Round Well, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-LB0109 4TI-0110 4TI-0116 4TI-0117
76638-126CS 220.54 USD
76638-126 76638-138 76638-226 76638-060
Storage Microplates, 96 Round Well, Azenta Life Sciences
Microplates Deep Well Microplates
These shallow 96 well storage microplates are particularly suitable for collection and preservation of samples widely used in cell biology research, molecular biology research, and drug discovery. Clear plate, perfect for storage applications.
  • Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
  • Autoclavable and stackable
  • Free from RNase, DNase, human genomic DNA and endotoxins
  • Alphanumeric grid reference to aid well and sample identification
  • SBS footprint
  • Round wells, suitable for most applications as they reduce droplet effects and wicking
  • Temperature range for use: –80 to 120 °C
  • Suitable for adhesive and heat sealing, and as a collection plate from filter systems
Low binding option made of selected low bind polymers, no coating is used to achieve the binding characteristics; they feature stacking ribs under the deck for improved stability, strengthening the plate for use in robotic automation applications.
V-Shaped bottom is best for precipitation, centrifugation and small volume recovery due to the V-shaped bottom, as the liquid gathers at the lowest point of the V for easy pipetting.
U-shaped bottom plates are best for washing, mixing and pelleting and give high surface area.
Ultra-low DNA binding option available (4ti-LB0109) for sensitive applications with ultra-low DNA input and for maximum DNA recovery after low temperature storage and high temperature incubation; learn more about our low binding range (not suitable for use on PCR blocks).
Accessories information: 96 round well sealing cap mat, clear silicone, for use with round 96 well microplates and deep well storage microplates.