PCR Plates, 384-Well, Fully Skirted, Roche Style, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-1381
76638-178CS 259.48 USD
76638-178
PCR Plates, 384-Well, Fully Skirted, Roche Style, Azenta Life Sciences
Microplates PCR Plates
These plates are designed for use on the Roche LightCycler 480 with 384 well block. Polypropylene, cut corner A24 and P24; working volume: <30 µl, total well capacity: 55 µl.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • 30 µl working volume, 55 µl total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with standard multichannel pipettes
The dimensions of these plates are designed for optimum compatibility with the Roche LightCycler® 480 and are in a 384 well format for reaction volumes of up to 30 µl.
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility, and as such comply with the same stringent requirements as the FrameStar® range.
Ordering information: Available barcoded upon request.