Storage Microplates, 96 Square Well, Deep Well, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0132 4TI-0136 4TI-0126
76638-174CS 430.06 USD
76638-174 76638-002 76638-056
Storage Microplates, 96 Square Well, Deep Well, Azenta Life Sciences
Microplates Deep Well Microplates
These storage plates are designed for high density sample collection and storage for a wide array of applications within cell biology, molecular biology and drug discovery. Clear plate, perfect for storage applications.
  • Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
  • Autoclavable and stackable
  • Free from RNase, DNase, human genomic DNA and endotoxins
  • Alphanumeric grid reference to aid well and sample identification
  • SBS footprint
  • Square wells, to make the best use of space, and to improve mixing
  • Temperature range for use: –80 to 120 °C
  • Suitable for adhesive and heat sealing, and as a collection plate from filter systems
V-Shaped bottom is best for precipitation, centrifugation and small volume recovery due to the V-shaped bottom, as the liquid gathers at the lowest point of the V for easy pipetting.
U-shaped bottom plates are best for washing, mixing and pelleting and give high surface area.
Accessories information: 96 square well sealing cap mat, clear silicone, for use with square 96 well microplates and deep well storage microplates.