PCR Foil Adhesive Seals, Peelable, Pierceable
Supplier: AZENTA US, INC
4TI-0550
76637-938CS 129 USD
76637-938
PCR Foil Adhesive Seals, Peelable, Pierceable
Seals Microplate Seals
The PCR aluminum foil seal has a strong acrylic adhesive which produces a seal of high integrity.
  • Pierceable
  • Peelable
  • Perforated end tabs for easy application and removal by peeling
  • Can be cut to fit partial plates
  • Temperature range: –40 to +120 °C
  • Non sterile
Adhesive seals provide a versatile, yet robust, method for protecting samples. To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.
Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.