Heat Sealing Foils, Peelable, DMSO Resistant
Supplier: AZENTA US, INC
4TI-0586-SLR 4TI-0587 4TI-0585 4TI-0585/100 4TI-0585/S 4TI-0585-SLR 4TI-0586 4TI-0586/S
76637-694EA 934.21 USD
76637-694 76637-860 76637-726 76637-744 76637-814 76637-766 76637-894 76637-890
Heat Sealing Foils, Peelable, DMSO Resistant
Seals Microplate Seals
The DMSO resistant peel heat seal is a foil seal compatible with polypropylene and forming an excellent seal to cyclic olefin copolymer (COC) plates.
  • Permanent seal to polyethylene
  • Peelable seal to polypropylene and COC
  • High solvent resistance, including 100% DMSO
  • Suitable for long term storage
  • For low and room temperature compound storage in Dimethyl Sulfoxide (DMSO) and organic solvents
  • Temperature range: –80 to +40 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.