Heat Sealing Foils, Peelable, Universal Plate Compatability
Supplier: AZENTA US, INC
4TI-0523 4TI-0523/S 4TI-05231 4TI-0523-SLR 4TI-0524 4TI-0524/S 4TI-0524-SLR
76637-850EA 1393.46 USD
76637-850 76637-800 76637-732 76637-924 76637-884 76637-868 76637-698
Heat Sealing Foils, Peelable, Universal Plate Compatability
Seals Microplate Seals
The universal foil heat sealing peelable film is a laminate seal compatible with PP, PE, PS, COC and PC plates, providing the highest flexibility in plate material choice.
  • Suitable for low temperature sample storage and high temperature uses, such as PCR
  • Wide material compatibility allows for high throughput sealing of different plates without the need for roll changes
  • Resealable by applying another Universal Foil Heat Sealing Peelable Film directly on top of a previously pierced seal
  • Pierceable with needle, but not with standard pipette tips
  • Autoclavable (121 °C)
  • Temperature range: –80 to +90 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective. Universal foil heat sealing peelable film forms a complete seal to a plate enabling low temperature uses as well as high temperature uses, such as PCR (when used with a pressurized heated lid).
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers.