PCR Foil Adhesive Seals, Peelable, Pierceable, DMSO Resistant
Supplier: AZENTA US, INC
4TI-0512
76637-746CS 559.11 USD
76637-746
PCR Foil Adhesive Seals, Peelable, Pierceable, DMSO Resistant
Seals Microplate Seals
The PCR aluminum foil seal has a chemically resistant silicone adhesive layer to produce a seal with high levels of solvent resistance, including to Dimethyl Sulfoxide (DMSO).
  • Peelable
  • Perforated end tabs for easy application and removal by peeling
  • No sticky residue on the plate surface following removal
  • Temperature range: –20 to +80 °C
  • Non sterile
Adhesive seals provide a versatile, yet robust, method for protecting samples. To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.
Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.
Ordering information: Gamma treatment available upon request.