PCR Foil Adhesive Seal, Peelable, Pierceable, Strong Adhesive
Supplier: AZENTA US, INC
4TI-0500FL
76637-706CS 177.68 USD
76637-706
PCR Foil Adhesive Seal, Peelable, Pierceable, Strong Adhesive
Seals Microplate Seals
The PCR aluminum foil seal has a strong acrylic adhesive which produces a seal of high integrity.
  • Pierceable
  • Peelable
  • Perforated end tabs for easy application and removal by peeling
  • Temperature range: –80 to +110 °C
  • Non sterile
Adhesive seals provide a versatile, yet robust, method for protecting samples.
To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.
Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.