COSA™ Foam 40, Ecolab® Life Sciences
Supplier: ECOLAB LIFE SCIENCES
6301143
76637-528CS 780.02 USD
76637-528
COSA™ Foam 40, Ecolab® Life Sciences
Cleaning Agents Detergents and Surface Cleansers
COSA™ Foam 40 is a liquid, neutral pH product for manual cleaning, or for use as a foam additive in cleaning solutions with Cosa™ CIP products for the pharmaceutical and personal care industries.
  • Suitable for manual and foam cleaning
  • Clear solution at all temperatures
  • Low toxicity high LD50-value and PDE value
  • cGMP manufactured - Global formulation
Safe for manual use, COSA™ Foam 40 acts as a mild and odorless detergent.