Non-Coded Sample Storage Tubes, 0.3 ml, 96-format, Internal Thread, Azenta Life Sciences
Supplier: AZENTA US, INC
66-32040 66-32040-Y6 66-32041 66-32041-Y6 66-32041-Y6-L 66-32141
76637-078CS 65.86 USD
76637-078 76637-026 76636-668 76637-080 76636-878 76636-874
Non-Coded Sample Storage Tubes, 0.3 ml, 96-format, Internal Thread, Azenta Life Sciences
Vials
The same storage tubes are also available in non-coded format for independent customization.
  • Manufactured from high-quality virgin polypropylene
  • Suitable for cryogenic storage as low as –196 °C (not for use in liquid phase nitrogen)
  • Secure sample storage and tracking
Ordering information: Available bulk uncapped or capped.