Coriolis® Micro Air Sampler, Bertin Instruments
Supplier: BERTIN CORP
P001080-CORM0-A.0
76636-418EA 11698.84 USD
76636-418
Coriolis® Micro Air Sampler, Bertin Instruments
Samplers Air Samplers
Coriolis® Micro is a portable biological air sampler for indoor or outdoor air bio-contamination monitoring. The equipment concentrates airborne particles in a liquid sample that can be analysed afterwards using microbiological techniques.
  • Concentrates airborne particles in a liquid sample
  • Compatible with culture and molecular biology standard methods (qPCR included)
  • Easy decontamination (hydrogen peroxide)
  • Autonomy on battery of 1 hour
  • Optional longer collection system up to 6 hours
Sample types include toxins, virus, bacteria, moulds, pollens, and spores.
Suitable for environmental and pollution research, pharmaceutical, food and veterinary industries, biomedical and health environments.
Accessories information: Plastic cones are used with a screw cap to facilitate the transport and preservation of samples. Sterile and nonsterile cones and caps are available (ready to use doses).
The long-time monitoring option allows the user to maintain constant liquid volume over a longer collection period. External power source is required.