Whatman™ Quartz Air Sampling Filter Papers, Grade QM-C, Whatman products (Cytiva)
Supplier: Cytiva
1855-037 1855-047 1855-090 1855-150 1855-865 1855-866 1855-082 1855-110 1855-045 1855-050 1855-070 1855-085
76402-780PK 227.56 USD
76402-780 76402-782 76402-784 76402-786 76402-788 76402-790 76402-824 76402-826 76498-680 76498-682 76635-536 76635-528
Whatman™ Quartz Air Sampling Filter Papers, Grade QM-C, Whatman products (Cytiva)
Filters Glass Fiber Filters
QM-C 100% quartz microfiber filter paper features low trace metals and high temperature resistance. It is well-suited for heavy metal analysis of particulate matter and sampling of acidic gases, stacks, flues and aerosols in air monitors.
  • 100% pure quartz microfiber, without any binder or organic chemicals
  • Pre-fired filters eliminate firing step, saving time in prepartion process
  • Air retention efficiency ≥99.95% at 0.3 μm DOP
QM-C high purity quartz microfiber filter addresses the challenge of heavy metal analysis in PM2.5/PM10. It is also suitable for organic chemical analyses such as PCBs and PAHs.
Certifications: Certified to conform to European and US EPA standards for heavy metal analysis in air particulate matter.