SteelFlex™ Blue Nitrile Examination Gloves
Supplier: AMERICAN NITRILE OPERATIONS, LLC
ANST351 ANST352 ANST353 ANST354 ANST355
76635-430BX 20.81 USD
76635-430 76635-432 76635-434 76635-436 76635-438
SteelFlex™ Blue Nitrile Examination Gloves
Gloves
Made in the USA, these premium nitrile gloves are designed for use in medical and healthcare applications.
  • Enhanced sensitivity
  • Reduces allergens
  • 3.5 mil thickness
  • Superior 1.5 AQL
  • Complies with ASTM standards
  • Cleared for use with chemotherapy drugs in accordance with ASTM D6978
SteelFlex™ gloves provide a barrier that protects patients and practitioners alike, and allows for enhanced tactile sensitivity and dexterity to deliver optimal care.
Featuring compression fit technology and a latex-free and powder-free design, these glove are intended for medical and research purposes. They are ideally suited for use in medical, veterinary, laboratory, dental, and retail industries.