Anti-Uroplakin ll/lll cocktail Multiple Recombinant Antibody
Supplier: Diagnostic Biosystems
RPM001-01 RPM001-05 RPM001
76635-210EA 222.87 USD
76635-210 76635-212 76635-214
Anti-Uroplakin ll/lll cocktail Multiple Recombinant Antibody
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. In non-neoplastic mammalian urothelium, UPs are expressed in the luminal surface plasmalemma of superficial (umbrella) cells, Uroplakin II/III cocktail is specific for tumors of urothelial origin and, when used in combination with other markers, can aid in the diagnosis of primary and metastatic tumors.
  • Hosts: Mouse and Rabbit