Anti-Progesterone (PR) Rabbit Monoclonal Antibody [clone: SP2]
Supplier: Diagnostic Biosystems
RMAB002-01 RMAB002-05 RMAB002
76634-696EA 364.24 USD
76634-696 76634-698 76634-700
Anti-Progesterone (PR) Rabbit Monoclonal Antibody [clone: SP2]
Antibodies
The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR indicates a responsive estrogen receptor (ER) pathway. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.