Anti-HSV type I and II cocktail Multiple Recombinant Antibody
Supplier: Diagnostic Biosystems
PDRM001
76634-686EA 224.16 USD
76634-686
Anti-HSV type I and II cocktail Multiple Recombinant Antibody
Antibodies
This antibody reacts with HSV type I and II specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
  • Hosts: Mouse and Rabbit