Anti-CD57 HNK-1 Mouse Monoclonal Antibody [clone: NK-1]
Supplier: Diagnostic Biosystems
PDM130
76634-382EA 238.16 USD
76634-382
Anti-CD57 HNK-1 Mouse Monoclonal Antibody [clone: NK-1]
Antibodies
This antibody is specific to a 110 kDa human CD57 myeloid associated glycoprotein. It stains mononuclear cells with natural killer activity, neuroendocrine cells and their tumors.