Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Supplier: Diagnostic Biosystems
PDM127
76634-378EA 286.57 USD
76634-378
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Antibodies
This antibody reacts with a 220 kDa protein. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 is very useful in the identification of Hodgkin disease.