Anti-CD74 B cell Mouse Monoclonal Antibody [clone: LN2]
Supplier: Diagnostic Biosystems
PDM033
76634-292EA 417.72 USD
76634-292
Anti-CD74 B cell Mouse Monoclonal Antibody [clone: LN2]
Antibodies
This antibody is specific to CD74. This antibody can be used for the identification of B cell lymphomas and leukemias. It also stains activated neoplastic cells in T cell lymphomas and Sternberg-Reed cells.