Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]
Supplier: Diagnostic Biosystems
MOB565
76634-138EA 85 USD
76634-138 76634-140 76634-142
Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]
Antibodies
SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas, and benign tumors such as schwannomas.SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.