Magnetic Separation Racks, Cytiva
Supplier: Cytiva
29710714
76633-056EA 1000.33 USD
76633-056
Magnetic Separation Racks, Cytiva
Racks Magnetic Separation Racks
Magnetic separation racks containing a neodymium magnet for small-scale protein purification and sample enrichment with magnetic beads.
  • Strong magnetic field, providing quick and efficient separation of magnetic beads from various liquid sample media used routinely for nucleic acid extraction procedures
  • Easily remove the magnetic block from the plastic rack to allow rapid demagnetization of the samples
  • Process up to six samples in parallel with easy pipette access
The magnetic field is strong, providing quick and efficient separation of magnetic beads from various liquid sample media, including viscous samples (e.g., blood or plasma diluted in typical viscid lysis/binding buffers used routinely for nucleic acid extraction procedures).
The magnetic beads are quickly attracted to magnets located at the back of the Magnetic Separation Rack, so the bead pellet is distributed along just one side of the tube permitting maximum bead retention and yield. Easily remove the magnet block from the plastic rack to allow rapid demagnetization of the samples.
Complementary to the following magnetic bead-based products: SeraSil-Mag™ silica coated superparamagnetic particles, Sera-Mag™ SpeedBeads Carboxylate-Modified Magnetic Particles, or Sera-Mag™ Carboxylate-Modified Magnetic Particles.