Presto™ Laser Histology Microscope Slide Printer, Azer Scientific
Supplier: VWR International
76632-834
76632-834EA 40013.46 USD
76632-834
Presto™ Laser Histology Microscope Slide Printer, Azer Scientific
Printers Cassette and Slide Printers
Azer Presto™ Microscope Slide Printer allows for quick, permanent and efficient printing on microscope slides. UV laser technology eliminates the need of expensive printer consumables and maintenance.
  • Integrate to LIS/HIS system
  • High resolution 500×500 dpi
  • High speed; as fast as three seconds for on-demand printing
  • Input cartridge holds up to 100 slides and output holds up to 35 slides
  • Continuous and on-demanding printing model
  • Barcode, QR code, logo, alphanumeric, graphic printing
  • ≤65dB work noise
Azer Scientific's Presto™ microscope slide printer is an ultraviolet laser model that can integrate into any laboratory or hospital information system. Our UV laser technology creates a permanent labeling resistant to degradation. Implementing the new Azer Scientific Presto™ slide printer aids in a time management workflow that you will not want to resist.
Ordering information: Two year warranty.
Delivery information: Includes two pre-installed templates. Contact prior to shipping for data fields required.
Caution: Do not attempt to open during operation.