FrameStar® PCR Plates, 384-Well, Fully Skirted, Roche Style, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0380/C 4TI-0381 4TI-0382 4TI-0383
76629-096CS 307.46 USD
76629-096 76629-144 76629-098 76629-046
FrameStar® PCR Plates, 384-Well, Fully Skirted, Roche Style, Azenta Life Sciences
Microplates PCR Plates
Polypropylene wells, polycarbonate frame, cut corners A24 and P24; working volume: <30 µl, total well capacity: 55 µl. Ideal for use with robotic systems. Compatible with Roche LightCycler® 480 with 384 well block and standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for Improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
The dimensions of these plates are designed for optimum compatibility with the Roche LightCycler® 480, and are in a 384 well format for reaction volumes of up to 30 µl.
The rigid two-component design eliminates warping and distortion during the PCR process, making it ideal for use with robotic systems and the skirt allows for labeling or barcoding.
Ordering information: Available barcoded upon request.