Bacto™ Tryptose, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211709 211713
76628-898EA 5207.9 USD
76628-898 76628-952
Bacto™ Tryptose, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.
  • Reported as beneficial for cell culture applications
  • Animal origin (AO) nutritional supplement
  • Tryptose is very useful for cultivation of Streptococci, Pneumococci, Meningococci, and other Fastidious organisms
  • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production
For use in cell culture, Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.