Difco™ Tryptone Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211921 211922 211923
76628-804EA 164.2 USD
76628-804 76628-908 76628-934
Difco™ Tryptone Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products.
  • Animal origin, pancreatic digest of casein
  • Primary nitrogen source of trypticase soy broth and agar
  • Referenced in Official Methods of Analysis of AOAC International
  • Supports the growth of many different types of bacteria and fungi
  • Recommended for use in media formulations when good growth of fungi and bacteria is required
  • Used in the development of animal health vaccines and with human health Botox-like products
Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production. Customers have used this supplement in the development of animal health vaccines, such as Clostridial vaccines, and with human health Botox-like products produced from Clostridium botulinum.
Certifications: LUO