CHO Medium, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670006
76628-902EA 9544.4 USD
76628-902
CHO Medium, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Prepared Media for Microbiology
CHO Medium is a high-performance, complete, animal origin-free (AOF) formulation offering high-production yields without the need for further supplementation. This medium is formulated to support cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures.
  • Chemically defined (CD)
  • Protein-free
  • Serum-free
CHO Medium can be used in various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications.