Bacto™ Yeast Extract, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
212710 212720 212730 212750
76628-840EA 13716.35 USD
76628-840 76628-870 76628-866 76628-828
Bacto™ Yeast Extract, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. They are derived from primary grown baker’s yeast. These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications.
Related yeast extract products:
Gibco Yeast Extract - developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
Difco Yeast Extract UF - ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin levels and high content of naturally occurring B vitamins, it can be used to effectively reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
Difco Yeast Extract Low-Dusting (LD) - designed to limit the amount of dust generated compared to other spray-dried yeast extracts.
Bacto Yeast Extract is one of the most complete and versatile fermentation supplements available. It is an important ingredient for the microbiological assay of vitamins. Yeast extract is also of value in the assay of antibiotics. B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract. Bacto Yeast Extract, Technical, was developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics.