Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211692 211693 212220 212230
76628-928EA 46979.3 USD
76628-928 76628-922 76628-782 76628-858
Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
Bacto Proteose Peptone No. 4 offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
Certifications: LUO, MUO.