BiTek™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
253720
76628-754EA
8805.28
USD
76628-754
BiTek™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology
Dehydrated Media for Microbiology
BiTek Proteose Peptone No.3 was developed to provide an alternative product to Bacto Proteose Peptone No.3, with similar growth characteristics.
- Enzymatic digest of protein
- Used in preparing microbiological and mammalian cell culture media
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)